VWR® TotalBLOT+™ , Blotting membranes, PVDF
供应商: Avantor
VWRCE578-1ROLLEA 15380 CNY
VWRCE578-1ROLL
VWR® TotalBLOT+™ , Blotting membranes, PVDF
TotalBLOT+™ PVDF membranes are naturally hydrophobic membranes which exhibit strong hydrophobic interactions with a wide range of proteins.
  • Chemically resistant and does not shrink or disfigure during destaining in methanol
  • Pure white microporous solid phase support
  • Allows sensitive immuno-detection down to nanogram or picogram levels
  • Direct visualisation of immobilised proteins with all common staining reagents, including Coomassie Blue, Amido Black, and Ponceau S
  • Pore size: 0,2 µm
TotalBLOT+™ PVDF membranes are ideal for Western transfers, protein binding assays and membrane bound protein sequencing. In addition, resistance to trichloroacetic acid and triethylamine allows direct insertion of the membrane into amino acid analysers and gas phase protein sequencing systems.