Master mix, Extender™ PCR-To-Gel, 2X
供应商: VWR Life Science
VWRCN867-2X1.25MLEA 2379.08 CNY
VWRCN867-2X1.25ML
Master mix, Extender™ PCR-To-Gel, 2X
Extender™ PCR-to-Gel Master Mix, 2X offers a single-step procedure for performing PCR reactions followed by analysis on agarose gels without the addition of loading buffer. The master mix includes Extender™ Taq Polymerase, dNTPs, MgCl2 and tracking dyes. The user supplies primer and template DNA. Aliquots of the PCR reaction can be directly loaded onto an agarose gel.
  • Pre-mixed formulation saves time
  • Streamlines PCR sample handling and processing
  • Includes all reagents expect fluorescent DNA dye, DNA template and primer DNA