Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
China
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
¥0.00
Order Entry
Home
Proteins and Peptides
Bovine serum albumin (BSA), Fraction V
Bovine serum albumin (BSA), Fraction V
Bovine serum albumin (BSA), Fraction V
:
Thermo Fisher Scientific
Bovine serum albumin (BSA), Fraction V
:
Thermo Fisher Scientific
White to yellow to light beige Crystalline powder
Source:
Species:
Environmentally Preferable:
ACRO13473
Pack type
Pk
ACRO134730100
134730100
Glass bottle
10 g
1 * 10 克
1 * 10 克 - ¥0.00
¥0.00
-
+
1 * 10 克
¥0.00
-
+
ACRO134731000
134731000
Glass bottle
100 g
1 * 100 克
1 * 100 克 - ¥0.00
¥0.00
-
+
1 * 100 克
¥0.00
-
+