Fmoc-Lys(Boc)-OH 98%

供应商: Thermo Scientific

Synonyms: Nα-芴甲氧羰基-Nε-叔丁氧羰基-L-赖氨酸

ACRO265220010EA 0 CNY
ACRO265220250 ACRO265220100 ACRO265220010
Fmoc-Lys(Boc)-OH 98%
Fmoc-Lys(Boc)-OH
分子式: C₂₆H₃₂N₂O₆
分子量: 468,55 g/mol
溶点: 129…135 °C
存储条件: Refrigerator
MDL编号: MFCD00037138
CAS号: 71989-26-9

现在订购

产品技术规格

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -11° to -13° (20°C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR