氯甲酸异丙烯酯 96% stabilised

供应商: Thermo Scientific
Danger

ACRO275120050EA 0 CNY
ACRO275120050 ACRO275120010 ACRO275120250
氯甲酸异丙烯酯 96% stabilised
氯甲酸异丙烯酯
分子式: C₄H₅ClO₂
分子量: 120,54 g/mol
沸点: 95…97 °C (760 mmHg)
密度: 1,103 g/cm³
闪点: 10 °C
存储条件: Refrigerator
MDL编号: MFCD00037083
CAS号: 57933-83-2
UN号: 3489
ADR: 6.1,I

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4160 (20°C, 589 nm)
GC ≥95 %
Stabilizer Approx. 0.01% BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR