Tetrodotoxin 99%

Danger

ACRO328560010EA 0 CNY
ACRO328560010
Tetrodotoxin 99%
Tetrodotoxin
分子式: C₁₁H₁₇N₃O₈
分子量: 319,27 g/mol
存储条件: Refrigerator
MDL编号: MFCD00213719
CAS号: 4368-28-9
UN号: 3462
ADR: 6.1,I

现在订购

产品技术规格

Appearance White to off-white Powder or solid or film
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR