Fumonisin B2 96+%

供应商: Thermo Scientific
Danger

ACRO328260010EA 0 CNY
ACRO328260010
Fumonisin B2 96+%
Fumonisin B2
分子式: C₃₄H₅₉NO₁₄
分子量: 705,839 g/mol
存储条件: Freezer
MDL编号: MFCD00133350
CAS号: 116355-84-1
UN号: 2811
ADR: 6.1,I

现在订购

产品技术规格

Appearance White to light yellow to brown Solid
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR