双(叔丁基环戊二烯)二氯化钛(IV)

供应商: Thermo Scientific
Warning

ACRO347860010EA 0 CNY
ACRO347860010
双(叔丁基环戊二烯)二氯化钛(IV)
双(叔丁基环戊二烯)二氯化钛(IV)
分子式: C₁₈H₂₆Cl₂Ti
分子量: 361,172 g/mol
溶点: 233 °C
存储条件: Ambient
MDL编号: MFCD01073774
CAS号: 79269-71-9

现在订购

产品技术规格

Appearance Bordeaux Crystals
Infrared spectrum Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR