TentaGel® S-RAM
供应商: Thermo Scientific产品技术规格
Appearance | White to yellow Powder |
Particle size | 90 µm |
Loading | 0.2 to 0.3 mmol/g |
Learn more
About VWR
Appearance | White to yellow Powder |
Particle size | 90 µm |
Loading | 0.2 to 0.3 mmol/g |
© 2024 VWR International, LLC. All rights reserved.
© 2024 FORTUNE Media IP Limited All rights reserved. Used under license.