4-Amino-4-carboxytetrahydrothiopyran hydrochloride 95%

供应商: Thermo Scientific
Warning

ACRO360505000EA 0 CNY
ACRO360505000
4-Amino-4-carboxytetrahydrothiopyran hydrochloride 95%
4-Amino-4-carboxytetrahydrothiopyran hydrochloride
分子式: C₆H₁₂ClNO₂S
分子量: 197,677 g/mol
存储条件: Refrigerator
MDL编号: MFCD02683141
CAS号: 67639-41-2

现在订购

产品技术规格

Appearance Grey-brown Crystalline powder
Infrared spectrum Conforms
NMR ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR