3,4-Di-O-acetyl-L-arabinal 97%

供应商: Acros Organics

ACRO370380010EA 0 CNY
ACRO370380010
3,4-Di-O-acetyl-L-arabinal 97%
3,4-Di-O-acetyl-L-arabinal
分子式: C₉H₁₂O₅
存储条件: Ambient
MDL编号: MFCD04112729
CAS号: 3945-18-4

现在订购

产品技术规格

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR