N,N-Diformylacetamide 80%
供应商: Thermo Scientific
|
Warning
|
ACRO380450050EA
0
CNY
ACRO380450050
ACRO380450010
N,N-Diformylacetamide 80%
N,N-Diformylacetamide
分子式:
C₄H₅NO₃ 分子量: 115,088 g/mol 沸点: 70 °C (15 mmHg) 存储条件: Refrigerator |
MDL编号:
MFCD00015916 CAS号: 26944-31-0 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥75 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...