High perfomance data loggers, Squirrel 2040
供应商: Grant
GRANSQ2040-4F16EA 0 CNY
GRANSQ2040-4F16
High perfomance data loggers, Squirrel 2040
These loggers are commonly used to measure current, voltage, resistance and temperature. It has sixteen to thirty two analogue input channels. All can be logged or used as triggers to automatically start or stop logging.
  • Sixteen true differential or 32 single ended analogue inputs for voltage, current or resistance measurements plus two high voltage, four pulse and eight digital event/state inputs
  • Logging rates up to 100 Hz on up to four channels
  • Analogue inputs for use with thermistors, thermocouples, 2, 3 or 4 wire RTD temperature sensors and from 4 to 20 mA signals
  • Flexible communications and high speed option
This is a compact, portable logger which is also suitable for bench based and fixed installations. Easily programmed via the four integral push buttons and large graphical display and with a basic accuracy, the loggers are able to fulfil many routine data logging needs, including more demanding applications requiring up to 10 readings per second on one channel.
Twin processors, multiple 24-bit A to D converters, up to 32 universal channels, removable Multi Media (MMC) or SD card memory and a choice of communications methods ensure that the Squirrel 2040 series provides state-of-the-art data logging and communication capability for sophisticated applications needs. They are easy, fast and convenient to use – either as portable, stand-alone loggers or as PC-linked data acquisition systems in industrial testing and research, scientific and environmental research and quality assurance applications.
The Squirrel 2040 series provide high performance universal data loggers packed with the same powerful features as the Squirrel 2020 series with additional high speed logging on up to four channels and twice as many universal input channels. This provides great flexibility to handle a wide range of complex and demanding multi-channel applications such as high-speed vehicle testing or engine monitoring. Flexible communications (USB, Ethernet, RS232).
Delivery information: Supplied with software, USB connection cable, batteries, 4 Precision Resistors for 4 - 20 mA connection.