Mobile frame, FG502
供应商: Bochem
BOCHFG502EA 0 CNY
BOCHFG502
Mobile frame, FG502
Mobile frames made of stainless steel.
  • Complete setup on rolls
  • Cage structure: Encloses the whole apparatus
  • Fixation of frame construction at side or back boards
  • Relocatable fixation tubes for side or back boards (min. depth: 750 mm)
  • Incl. stainless steel collecting tray for leackages (easy to take off)