dNTP mix, 25 mM solution
供应商: VWR Chemicals
VWR®
VWRCE636-40UMOLEEA 810.33 CNY
VWRCE636-40UMOLE
dNTP mix, 25 mM solution
核酸试剂 核苷酸
Ready to use dNTP mix. One tube of 10 μmol of each dNTP in a 25 mM solution, pH 7,0. Tube volume 400 μl.
  • Building blocks for the amplification of DNA
  • High purity grade