Hydrocarbon oil index determination standards in Certan bottles according to ISO 93772-2
供应商: VWR Chemicals
VWRC87503.002EA 4826 CNY
VWRC87503.002 VWRC87502.004 87502.002 VWRC87505.004 VWRC87502.003 VWRC87504.002 VWRC87505.003 VWRC87503.001 VWRC87504.003 VWRC87504.001 VWRC87504.004 VWRC87505.002 VWRC87502.001 VWRC87503.003 VWRC87503.004 VWRC87505.001
Hydrocarbon oil index determination standards in Certan bottles according to ISO 93772-2
标准品 环境分析标准
Designed for analysis of water and ground samples, according to the ISO 9377-2 norm.
  • Delivered with certificate of analysis, reference and batch number of source materials
  • Packed in 1,5 and 4,5 Certan® bottles
Certan® bottles provide the advantage of a sealed ampoule with the flexibility of a screw cap bottle or a septum vial. These unique, screw-cap sample containers with a 1,2×28 mm height capillary opening are designed for the storage of volatile samples and reference standards.