Order Entry
VWR® ARISTAR®, Calcium Standard Solutions for ICP-MS
VWR® ARISTAR®, Calcium Standard Solutions for ICP-MS
undefined
VWR® ARISTAR®, Calcium Standard Solutions for ICP-MS
Supplier:  VWR Chemicals
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Environmentally Preferable:
Specifications
Product Information
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
  • VWRC457042S
    Calcium
    Ca
    10000 mg/L
    2-5% HNO₃
    Plastic bottle
    100 mL
    1 * 100 毫升
    1 * 100 毫升 - ¥3,663.00
    ¥3,663.00
    1 * 100 毫升
       ¥3,663.00
  • VWRC456692T
    Calcium
    Ca
    1000 mg/L
    2-5% HNO₃
    Plastic bottle
    100 mL
    1 * 100 毫升
    1 * 100 毫升 - ¥2,169.00
    ¥2,169.00
    1 * 100 毫升
       ¥2,169.00
  • VWRC85555.180
    Calcium
    Ca
    10 mg/L
    2-5% HNO₃
    Plastic bottle
    100 mL
    1 * 100 毫升
    1 * 100 毫升 - ¥1,301.00
    ¥1,301.00
    1 * 100 毫升
       ¥1,301.00