Anti-IgG Goat Polyclonal Antibody (Peroxidase)

供应商: Jackson Immunoresearch Laboratories
JACK101-035-008EA 0 CNY
JACK101-035-008
Anti-IgG Goat Polyclonal Antibody (Peroxidase)
抗体
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Peroxidase
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR