Anti-IgG Goat Polyclonal Antibody (Rhodamine Red-X)

供应商: Jackson Immunoresearch Laboratories
JACK109-035-098EA 0 CNY
JACK109-035-098
Anti-IgG Goat Polyclonal Antibody (Rhodamine Red-X)
抗体
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine Red-X
Epitope: F(c) γ
Host: Goat
Isotype: IgG
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR