Anti-FOXP3 Rat Antibody [clone: FJK-16s]

供应商: eBioscience
EBIO12-5773-82EA 0 CNY
EBIO12-5773-80 EBIO12-5773-82
Anti-FOXP3 Rat Antibody [clone: FJK-16s]
抗体
Type:
Antigen: FOXP3
Clonality:
Clone: FJK-16s
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Bovine, Dog, Cat, Mouse, Pig, Rat
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR