Anti-IgG Goat Antibody

供应商: Jackson Immunoresearch Laboratories
JACK115-005-062EA 0 CNY
JACK115-005-062
Anti-IgG Goat Antibody
抗体
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation:
Epitope: Heavy and Light Chain
Host: Goat
Isotype:
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR