Anti-LATS1/LATS2 Rabbit Polyclonal Antibody

供应商: BETHYL
BETHA300-479AEA 0 CNY
BETHA300-479A
Anti-LATS1/LATS2 Rabbit Polyclonal Antibody
抗体
Type: Primary
Antigen: LATS1/LATS2
Clonality: Polyclonal
Clone: Unconjugated
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR