dGTP, 100 mM solution, pH 7,0
VWRCE539-40UMEA 1079.15 CNY
VWRCE539-40UM
dGTP, 100 mM solution, pH 7,0
dGTP, 100 mM solution in sterile, deionised water at pH 7,0 to maintain stability.  Prepared from monosodium salts of the free nucleotides. No excess sodium ions are used in buffering to reduce sodium ion mediated inhibition of DNA amplification.