Economy cable lockout
供应商: Brady
BRDY45352EA 415 CNY
BRDY45352
Economy cable lockout
安全锁
This cable lockout is incredibly versatile making it perfect for all unusual energy isolating devices that are difficult to lock out.
  • Six lock holes in handle are 7,14 mm in diameter to accommodate most lock shackles and hasps
  • Made of fiberglass reinforced PP
Delivery information: Economy cable lockout includes 3 m cable.