Cell culture plates, 96-well, for fluorescent and luminescent applications
供应商: Corning
Corning®
CORN3917EA 3875 CNY
CORN3917 CORN3903 CORN3603 CORN3610 CORN3362 CORN3916 CORN3904
Cell culture plates, 96-well, for fluorescent and luminescent applications
微孔板 细胞培养板 Standard Plates
PS.
  • Black plates are designed to lower background in fluorescent assays
  • White plates are designed for luminescent assays
  • Treated for optimal cell attachment
  • Sterilised by gamma irradiation
  • Certified non pyrogenic