VWR® VisiLight® CL150, 卤素冷光源,
供应商: VWR
VWR® VisiLight®
VWRI631-1772EA 0 CNY
VWRI631-1772
VWR® VisiLight® CL150, 卤素冷光源,
冷光源
VisiLight®卤素冷光源CL150拥有模块化和灵活性的特点,适用于多种研究和工业应用。
  • 低噪制冷系统和无振动操作
  • 3270 K色温的150 W卤素灯泡
  • 采用现代设计、轻量、结实的铝质外壳
  • 0到100%可变光线强度,带电子调光和机械光圈
  • 日光(5400 K)、红色、绿色、蓝色和黄色滤光片选件
订购须知 Supplied without light guides, ring lights, focus optics or filters which must all be ordered separately.