Solid black and white microplates, 96-well
供应商: Corning®
Corning® NBS™ Corning®
CORN3600EA 9550 CNY
CORN3600 CORN3789A CORN3912 CORN3922 CORN3915 CORN3925 CORN3605 CORN4591 CORN3650 CORN3991 CORN3990 CORN3792
Solid black and white microplates, 96-well
微孔板 免疫微孔板和条带
PS, without lid, non sterile. Available either with NBS™ non binding surface, ideal for reducing protein and nucleic acid binding at low concentrations, and for increasing assay signal-to-noise ratio, or with High Bind surface for immunological applications. Also available untreated for solution based assays.
Available either with NBS™ non binding surface, ideal for reducing protein and nucleic acid binding at low concentrations, and for increasing assay signal-to-noise ratio, or with High Bind surface for immunological applications. Also available untreated for solution based assays.