Qualitative filter papers, standard grades, grade 2 and 2V, Whatman™
供应商: Whatman products (Cytiva)
Whatman™
WHAT1202-125EA 0 CNY
WHAT1202-125 WHAT1202-185 WHAT1002-931 WHAT1002-110 WHAT1002-917 WHAT1002-240 WHAT1002-090 WHAT1002-125 WHAT1002-070 WHAT1202-400 WHAT1002-185 WHAT1002-929 WHAT1002-270 WHAT1202-320 WHAT1202-270 WHAT1202-385 WHAT1202-240 WHAT1002-917_L WHAT1202-150 WHAT1002-320 WHAT1002-055 WHAT1202-500 WHAT1002-150 WHAT1002-047 WHAT1002-042 WHAT1002-385
Qualitative filter papers, standard grades, grade 2 and 2V, Whatman™
过滤器 滤纸 Qualitative Filter Paper
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Slightly more retentive and more absorbent than grade 1. In addition to general filtration in the 8 μm particle size range, the extra absorbency is utilised, for example, to hold soil nutrient in plant growth trials.
  • Folded filters expose extra surface area, which enables a higher loading capacity and shorter filtration time
  • Folded filters are suitable for extracting samples from soil to support contaminant testing in the presence of weakly acidic solutions