Sterilisation wraps, Bio-Shield®
供应商: Cardinal
Bio-Shield®
CARD4015 CARD4036 CARD4012 CARD4030 CARD4040
Sterilisation wraps, Bio-Shield®
灭菌包装
These versatile non woven wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
  • Wraps feature sterilisation tapes that adhere firmly and remove easily
  • Superior bacterial barrier properties compared to linen
  • Repels fluids