Safety wash bottles, wide neck, colour coded
供应商: Chemglass
CGLACG-1130-11EA 125 CNY
CGLACG-1130-11 CGLACG-1130-15 CGLACG-1130-04 CGLACG-1130-01 CGLACG-1130-06 CGLACG-1130-08
Safety wash bottles, wide neck, colour coded
洗瓶
These low-density polyethylene dispensing bottles are break-resistant and include DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ information on their exterior.
  • Easy-to-read
  • Intended for short-term dispensing use
  • Wide mouth for easy, safe filling
Polypropylene nozzle assemblies are leakproof and colour-coded to match bottle label. Toluene bottle is solid red with white lettering and hazard information.