Adapters for filter crucibles
供应商: Chemglass
CGLACG-1401-01EA 0 CNY
CGLACG-1401-01 CGLACG-1401-15
Adapters for filter crucibles
过滤器支架
Neoprene filter adapters are designed to fit between the filtering funnel and flask. For use with buchner funnels, grooch crucibles, filter tubes, etc. Eliminates the boring of rubber stoppers and the difficulties of inserting and removing glass stems in stoppers.
  • Can be used individually or nesting with adjacent sizes