Desiccator plate, porcelain
供应商: Chemglass
CGLACG8260230EA 0 CNY
CGLACG8260230
Desiccator plate, porcelain
干燥器 板式干燥器
Perforated porcelain desiccator plate with a large hole in the center for easy placement or removal from a glass dessicator and widely used in chemistry laboratories.
  • Plate is glazed on one side
  • High resistance to thermal shock
  • Very low expansion coefficient
  • High resistance to all reagents except hydrofluoric acid