Dewar, flask, stainless steel
供应商: Chemglass
CGLACG-1599-B-01EA 13196.52 CNY
CGLACG-1599-B-01
Dewar, flask, stainless steel
低温储存容器
Unbreakable dewar flask made entirely of stainless steel. Has flexible laboratory applications including cooling of small samples and as an insulating flask. Used for storage and transport of liquid nitrogen, dry ice CO₂ and other coolants.
Temperature range of −200 to +200 °C.