Volumetric flasks, class A, glass, clear, NS glass stopper
供应商: Chemglass
CGLACG-1600-08EA 0 CNY
CGLACG-1600-08
Volumetric flasks, class A, glass, clear, NS glass stopper
烧瓶 容量瓶
Flasks are manufactured to conform to Federal Specifications NNN-F-289 for Type 1, Class 'A' unserialized flasks and ASTM E-288.
  • Very good chemical resistance
  • Very good temperature resistance
  • Complete with solid glass pennyhead stopper