鞋套,PP
供应商: VWR, part of Avantor
VWR®
VWRI113-8283EA 0 CNY
VWRI113-8283 VWRI113-9052 VWRI113-8282
鞋套,PP
Disposable Garments
非编织PP,带CPE夹层鞋底。
  • 脚下无接缝
长×高:410×170 mm
Weight: 40 g/m² (SPP), 80 g/m² (CPE sole)
*通用尺寸
Packaging: 5 pieces per bundle, 10 bundles per bag, 4 bags per case.
*通用尺寸