Process supplement, HyClone Cell Boost™ 5
供应商: HyClone products (Cytiva)
HYCLSH30865.03EA 0 CNY
HYCLSH30865.01 HYCLSH30865.03
Process supplement, HyClone Cell Boost™ 5
细胞培养添加剂
HyClone™ Cell Boost™ 5 Supplement is designed to provide nutrients such as lipids, amino acids, vitamins, and growth factors as part of fed-batch cell culture strategies.
  • Chemically defined, protein-free and free of animal-derived components (ADCF)
  • Tested on a variety of cell lines, including Chinese hamster ovary (CHO), hybridoma, NS0, and human embryonic kidney (HEK) 293 cells
  • No L-glutamine or poloxamer 188
  • Ideal for supplementing nutrients for bioprocessing and media customisation applications
  • Manufactured according to cGMP guidelines