DNA extraction solution, QuickExtract™
供应商: Epicentre
EPICQE09050EA 0 CNY
EPICQE09050
DNA extraction solution, QuickExtract™
核酸试剂 核酸纯化试剂盒和试剂
The QuickExtract™ DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from almost any sample type using a simple, one-tube protocol that takes only 3 to 8 minutes, depending on the sample. QuickExtract Solution has been used to extract DNA from samples such as hair follicles, quill-end cells of feathers, tissue-culture cells, buccal cells, zebrafish organs and scales, and mouse tail snips. The extracted DNA is suitable for PCR analysis, such as genomic, transgenic, or viral DNA screening in animals, or for genetic or environmental research and screening in humans and other organisms.
The QuickExtract™ method allows for the processing of one to hundreds of samples simultaneously, without centrifugation, spin columns or the use of any toxic organic solvent. The method is also compatible with robotic automation.
  • Non-toxic reagents, inexpensive processing
  • Short procedure (8 minutes for the average sample)
  • No centrifugation or spin columns to reduce yields
  • Compatible with high-throughput and robotic workflows
订购须知 Supplied as a bulk solution, sufficient to perform 100×500 µl extractions.