Cover glasses, VistaVision™
供应商: VWR Collection
VWR® VistaVision™
VWRU16004-300EA 204 CNY
VWRU16004-320 VWRU16004-338 VWRU16004-300 VWRU16004-310
Cover glasses, VistaVision™
盖玻片
  • Resistant to chemicals
  • Free of bubbles amd scratches
  • No disturbing self-fluorescence under UV light
Certifications: ISO 8255-1