Immobilised polysaccharide columns
供应商: CHIRAL TECHNOLOGIES
CHIRALPAK® CHIRALCEL®
DAIC83524EA 0 CNY
DAIC87U82 DAIC81311 DAIC83335 DAIC81S84 DAIC81U82 DAIC85G24 DAIC81594 DAIC83322 DAIC84S83 DAIC81524 DAIC84324 DAIC84323 DAIC84335 DAIC83U83 DAIC84523 DAIC88G25 DAIC86G35 DAIC80S82 DAIC80335 DAIC80594 DAIC88525 DAIC85525 DAIC80G25 DAIC81325 DAIC84435 DAIC88592 DAIC88335 DAIC84524 DAIC81323 DAIC88225 DAIC85G35 DAIC83G35 DAIC88G35 DAIC83423 DAIC85G45 DAIC83524 DAIC88G45 DAIC86S84 DAIC88337 DAIC85511 DAIC83S84 DAIC83G45 DAIC80224 DAIC85S82 DAIC81U83 DAIC83226 DAIC80S84 DAIC88311 DAIC80525 DAIC87435 DAIC85423 DAIC80311 DAIC83S82 DAIC88435 DAIC88394 DAIC88324 DAIC83522 DAIC85S84 DAIC83U82 DAIC88323 DAIC80511 DAIC84G25 DAIC84311 DAIC80G35 DAIC85324 DAIC85435 DAIC81523 DAIC80345 DAIC87445 DAIC85311 DAIC85345 DAIC85394 DAIC88322 DAIC85524 DAIC88593 DAIC81S83 DAIC83311 DAIC85G25 DAIC84445 DAIC80226 DAIC81511 DAIC81324 DAIC88S82 DAIC84S84 DAIC87G35 DAIC85522 DAIC84G45 DAIC84594 DAIC85445 DAIC86G45 DAIC80337 DAIC85S83 DAIC86G25 DAIC87U83 DAIC83525 DAIC88224 DAIC84525 DAIC81322 DAIC83345 DAIC82224 DAIC88522 DAIC83325 DAIC88325 DAIC81335 DAIC84423 DAIC80225 DAIC88523 DAIC82225 DAIC84511 DAIC81423 DAIC83324 DAIC88445 DAIC85523 DAIC83G24 DAIC88G24 DAIC84325 DAIC80S83 DAIC88226 DAIC88595 DAIC84322 DAIC85325 DAIC80325 DAIC81337 DAIC84G24 DAIC87G25 DAIC80435 DAIC84522 DAIC83225 DAIC88524 DAIC80G45 DAIC83323 DAIC88594 DAIC88S84 DAIC87G24 DAIC87G45 DAIC80423 DAIC81345 DAIC84394 DAIC84S82 DAIC80322 DAIC80324 DAIC85323 DAIC86S82 DAIC84337 DAIC80523 DAIC83S83 DAIC86S83 DAIC85335 DAIC81525 DAIC81522 DAIC83523 DAIC80522 DAIC88423 DAIC85337 DAIC84G35 DAIC80445 DAIC83511 DAIC81S82 DAIC83224 DAIC86G24 DAIC80323 DAIC88511 DAIC88S83 DAIC85594 DAIC84345 DAIC87423 DAIC83594 DAIC88345 DAIC82226 DAIC83337 DAIC80G24 DAIC80524 DAIC83G25 DAIC85322
Immobilised polysaccharide columns
色谱柱
These columns allow chromatographers to employ essentially any organic solvent as mobile phase to develop methods for the most challenging separations. These columns are available in a variety of particle sizes and offer great speed and column resolving power. Columns packed with 3 µm CSPs offer high speed separations and a reduction in analysis time and cost per sample. Columns, packed with 3 µm particles, may be used in conventional HPLC units without significant loss in performance, even with the smaller column dimensions.
The immobilised columns are used for applications ranging from analytical method development to large-scale enantiomer resolutions.