Snap-on circuit breaker lockout
供应商: Brady
BRDY65387EA 67 CNY
BRDY65387
Snap-on circuit breaker lockout
安全锁
This snap-on breaker lockout is designed for 120 V circuit breakers that have holes in the switch tongue.
  • Easy and quick to install, just snap in place and apply lock
  • Device accepts lock shackles up to ⁹/₃₂" in diameter