Spreaders, T-shaped
供应商: COPAN Diagnostics
COPACOPTS-10EA 0 CNY
COPACOPTS-1 COPACOPTS-5 COPACOPTS-10
Spreaders, T-shaped
涂布棒 细胞涂布棒
These plastic T–shaped spreaders are designed for spreading and dispersing liquids across the surface of agar culture plates. T–shaped spreaders are used for performing bacterial cfu plate counts on water, milk, urine and other liquid samples.
  • Smooth rounded surfaces to prevent cutting or gouging of agar during inoculation
  • T–shape design ensures that even pressure is applied across the entire length of the T–bar in contact with the agar surface
Packaging: Available in three sterile configurations, as single wrapped, five spreaders per pack, or ten spreaders per pack.