Cut resistant gloves, Whizard®
供应商: Wells Lamont
Whizard®
WELLCM030004EA 0 CNY
WELLCM030004
Cut resistant gloves, Whizard®
手套
This ergonomically designed glove made from stainless steel provides superior cut and puncture protection.
  • Self-adjusting tensioning system on wrist allows for an easy fit with no straps or buckles to fasten
  • All Whizard® mesh gloves come with a metal serial number tag, which helps with reissuance and sizing information
  • Micro plasma welded rings have no gaps to trap soil that can feed bacterial growth
  • Made with 4 mm ring using 0,5 mm wire
  • Ambidextrous – turn inside out for the other hand
  • Easy to clean