PCR nucleotide mix
供应商: Promega
PROMC1141EA 0 CNY
PROMC1141 PROMU1432 PROMC1145 PROMU1431
PCR nucleotide mix
核酸试剂 核苷酸
PCR Nucleotide Mix is a premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP, each at a concentration of 10mM or 25mM in water.
  • cGMP-Grade Solution of dNTPs Pretested in PCR
  • cGMP-manufactured for lot-to-lot product consistency
  • Equimolar amounts of each dNTP in water
  • Two concentrations available: 10mM and 25mM
High-quality deoxynucleotide triphosphates (dNTPs) are critical for PCR success. PCR Nucleotide Mix is a cGMP-grade premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP. There are two formulations available: A premixed solution containing each nucleotide at a concentration of 10mM in water at pH 7.5; the total concentration of nucleotides is 40mM. A premixed solution containing each nucleotide at a concentration of 25mM in water at pH 7.5; the total concentration of nucleotides is 100mM. These PCR nucleotide mixes are ready to use and optimized for use in PCR.