The ergonomically designed MICROMAN® E positive-displacement pipettes, along with disposable capillary piston (CP) tips, provide the highest precision when pipetting viscous, non-aqueous liquids such as oils, blood, glycerol, and more while protecting the pipette from contamination.
- Reliable results even with non-aqueous liquids
- Overall protection
- Designed with comfort in mind
Contaminated, corrosive, or radioactive samples can generate aerosols that can cross-contaminate or damage the pipette. The disposable piston within the capillary piston (CP) tip is in direct contact with the liquid, preventing contamination and protecting the sample, the user, and the pipette. The unique QuickSnap system secures the disposable CP in one easy motion, fitting a CP as simply and reproducibly as a standard pipette. Easily eject the CP by pressing the push button and avoiding any contact with the disposable CP. The volume-control switch allows for easy volume adjustment and minimizes the risk of accidentally changing the volume during pipetting tasks.
Related Information
Capillary Micropipettes, MICROMAN® E
Application Notes
Gilson Achieve Pipetting Precision of Problem Liquids Gilson Exact and Secure Pipetting of Plasma and Whole Blood Gilson Improve HPLC Analytics with Proper Pipetting of Difficult Liquids Gilson Improve Pipetting of Viscous Liquids in Pharmaceutical Formulations Gilson Positive Displacement Pipette
Product Literature
TDS Positive Displacement Pipette
Datasheet
Product Information
- Catalog #Supplier #VolumeAccuracyImprecisionColour codePkAvailabilityPrice
- M10EM10EGILSFD10001FD100011 - 10 µl±0,09 - ±0,15 µl≤0,03 - ≤0,06 µlRed11 * 1 件¥0.00Specifications:
Accessories
Specifications
Environmentally Preferable - M25EM25EGILSFD10002FD100023 - 25 µl±0,25 - ±0,30 µl≤0,08 - ≤0,10 µlRed11 * 1 件¥0.00Specifications:
Accessories
Specifications
Environmentally Preferable - M50EM50EGILSFD10003FD1000320 - 50 µl±0,34 - ±0,70 µl≤0,20 - ≤0,30 µlBlue11 * 1 件¥0.00Specifications:
Accessories
Specifications
Environmentally Preferable - M100EM100EGILSFD10004FD1000410 - 100 µl±0,50 - ±1,00 µl≤0,20 - ≤0,40 µlBeige11 * 1 件¥0.00Specifications:
Accessories
Specifications
Environmentally Preferable - M250EM250EGILSFD10005FD1000550 - 250 µl±1,50 - ±2,50 µl≤0,30 - ≤0,50 µlRed11 * 1 件¥0.00Specifications:
Accessories
Specifications
Environmentally Preferable - M1000EM1000EGILSFD10006FD10006100 - 1000 µl±3,0 - ±8,0 µl≤1,6 - ≤4,0 µlLight blue11 * 1 件¥0.00Specifications:
Accessories
Specifications
Environmentally Preferable
Tips
Product Information
- Catalog #SeqTip styleVolumeSterileLengthFor use withPackedPkAvailabilityPrice