Cartridge filter, Durapore®
MILLCVDI01TPEEA 0 CNY
MILLCVDI01TPE
Cartridge filter, Durapore®
Superior filters for controlled low bioburden levels in biopharmaceutical fluids.
  • Non-fiber releasing
  • Ideal for LVP bulk pharmaceutical product filtration
  • Hydrophilic
Applicable for fine filtration of deionised water, in vitro diagnostics, oral suspensions, bulk pharmaceutical solutions and for media prefiltration.