J.T.Baker® BAKERBOND spe™ C18 Light Load, SPE Columns
供应商: Avantor
J.T.Baker® BAKERBOND spe™
BAKR7189-03EA 6457 CNY
BAKR7189-03
J.T.Baker® BAKERBOND spe™ C18 Light Load, SPE Columns
色谱柱
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Non ionic, non polar to polar analytes.
These SPE columns can be used in reversed phase mode.