SPE columns, J.T.Baker®, BAKERBOND spe™ C2
供应商: Avantor®
J.T.Baker®
BAKR7273-02
SPE columns, J.T.Baker®, BAKERBOND spe™ C2
色谱柱
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Polar and basic analytes.
These SPE columns can be used in reversed phase mode.