Jars, cloud and pour point, PYREX®, CORNING®
供应商: Corning
PYREX® Corning®
CORN6902EA 0 CNY
CORN6902
Jars, cloud and pour point, PYREX®, CORNING®
通用罐
Made of PYREX® glass, clear.
  • Not intended for vacuum use
  • Good chemical resistance
  • Good temperature resistance
These jars are used to measure the cloud point temperature in petroleum oils. A single line encircles the jar at a point approximately 51 mm above the bottom to assist in measuring the sample.
Certifications: Meets ASTM D-97, D-1500, D-2500 specifications.