Bomb for gum stability, PYREX®
供应商: Corning
PYREX®
CORN7732EA 0 CNY
CORN7732
Bomb for gum stability, PYREX®
石油试验装置
Bomb with cover made from PYREX® borosilicate glass to determine gum stability in gasoline.
  • Wide mouth with pourout style top
  • Cover to prevent condensed vapours from contaminating the sample
  • No interference with the free flow of oxygen
Replacement covers are available seperately.
Certifications: ASTM D-525