Cut resistant gloves, HyFlex® 11-625 / 11-627 / 11-628
供应商: Ansell
HyFlex®
ANSE11-625/7EA 0 CNY
ANSE11-627/10 ANSE11-625/7 ANSE11-625/10 ANSE11-627/7 ANSE11-628/7 ANSE11-628/8
Cut resistant gloves, HyFlex® 11-625 / 11-627 / 11-628
手套
The gloves are designed for use in dry, as well as slightly oily environments. The nylon/Lycra® liners have an innovative yarn structure that contains an optimal rate of Dyneema®, providing high levels of cut resistance.
  • Palm coated with polyurethane for additional protection
  • Lycra® liner and polyurethane coating provide excellent flexibility and fit
  • Elasticated cuffs
Ideal for contact with sharp pieces, pre-assembling, cutting of small dry or lightly oiled parts and maintenance applications.
Certifications: EN 388:2016 (4342B); Cat. II